Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Bottles and Carboys
VapLock™ Solvent Waste Kits for Bottles and Carboys collect effluent from multiple sources while minimizing harmful solvent exhaust
Brand:  Cole-Parmer™ WAK38NP-10
 View more versions of this product
Product Code. 15864534
  • 631.00 € / Each
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Various cap sizes available to fit your solvent container
Services
None
Specifications
Specifications
Solvent Safety
Polypropylene
1 x 1/2 in. NPT(F), 3 x 1/4 in. NPT(F), 6 x 1/4 in.-28 UN(F)(F)
Solvent-Compatible Containers
Nalgene 38-430
Solvent Waste Kit