Learn More
Cole-Parmer™ VapLock™ Solvent Waste Kits for Jars and Pails
VapLock™ Solvent Waste Kits for Jars and Pails collect effluent from multiple sources while minimizing harmful solvent exhaust
Brand:  Cole-Parmer™ WAK100P-10
 View more versions of this product
Product Code. 15844534
  • 718.00 € / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Various cap sizes available to fit your solvent container
Services
None
Specifications
Specifications
Solvent Safety
Polypropylene
1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F)
Solvent-Compatible Containers
100-415
Solvent Waste Kit