Learn More
Watson-Marlow 323 Series Drive with 313DW Pumphead
300 series cased pumps. Single or multi-channel benchtop pumps with manual, remote, analogue, RS232 control and accurate dispensing.
Brand:  Watson-Marlow 030.3124.3DE
 View more versions of this product
Product Code. 15306151
  • 3805.00 € / Each
Explore more special offers
Description
Description
  • 300 series laboratory pumps are a benchtop pumping solution with manual, remote, analogue or RS232 control capabilities. Featuring a chemically resistant ABS top and IP31 protection, 323E/D pumps have a membrane keypad that controls the pump, and a digital LCD to display speed. There are four models with increasing degrees of functionality:
  • 323E/DDu RS232/auto/manual enhanced-control high-flow pumps
  • 323E/DU and 323E/DU analogue-auto and enhanced manual-control pumps
  • 323E/DS high-performance manual control pumps
  • 323E/DE manual control pumps
Pumphead options available as standard (others available on request):
  • 313D and 313D2 three-roller pumpheads for continuous tubing
  • 314D and 314D2 four-roller pumpheads for continuous tubing
  • 314MC four-roller five-channel microcassette pumpheads
  • 318MC eight-roller five-channel microcassette pumpheads
  • 501RL and 501RL2 pumpheads for continuous tubing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Variable Speed
IP31
15 to 400 rpm
323E/D
Keypad, manual control
100/120/220/240 V
LCD
4.1 to 2000 mL/min.
Yes
313D and 313D2 pump heads for flow rates up to 2000mL/min; 314D and 314D2 for flow rates up to 1600mL/min
230 x 230 x 132 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Watson-Marlow 323 Series Drive with 313DW Pumphead > Model: 323E/D