Learn More
Eppendorf™ Varispenser™ 2, Bottle-Top Dispenser
Varispenser™ 2, Bottle-Top Dispenser is ideal for dispensing aliquots of liquid from large supply bottles.
330.00 € - 633.00 €
Specifications
Product Type Analog Bottletop Dispenser with Adapters
Channels 1
View More Specs
Products 6
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15707357
View Documents
Eppendorf™
4966000010
0.2 to 2 mL
330.00 €
Each
In Stock
to see stock.
Add to basket
15717357
View Documents
Eppendorf™
4966000029
0.5 to 5 mL
330.00 €
Each
In Stock
to see stock.
Add to basket
15727357
View Documents
Eppendorf™
4966000037
1 to 10 mL
343.37 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
15737357
View Documents
Eppendorf™
4966000045
2.5 to 25 mL
439.00 €
Each
In Stock
to see stock.
Add to basket
15747357
View Documents
Eppendorf™
4966000053
5.0 to 50 mL
508.53 €
Each
In Stock
to see stock.
Add to basket
15757357
View Documents
Eppendorf™
4966000061
10 to 100 mL
633.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Varispenser™ 2 is ideal for dispensing aliquots of liquid from large supply bottles.
  • Standard GL 45 thread on all sizes for most common bottle threads
  • Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
  • Screwable air vent cap for easy assembly of drying tube or microfilter
  • Oval-shaped dispenser housing for easy transportation
  • PFA seal of the sliding piston prevents seizing up
  • Fixation slide on interior toothed track for rapid and accurate volume adjustment
  • High chemical resistance of all dispensing components
  • Telescopic aspirating tubes for variable adjustment to bottle height
  • Fully autoclavable without disassembly
  • User adjustment option
Specifications
Specifications
Analog Bottletop Dispenser with Adapters
1
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ Varispenser™ 2, Bottle-Top Dispenser