Learn More
Eppendorf™ Varispenser™ 2, Bottle-Top Dispenser
Varispenser™ 2, Bottle-Top Dispenser is ideal for dispensing aliquots of liquid from large supply bottles.
Brand: Eppendorf™ 4966000045
433.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Varispenser™ 2 is ideal for dispensing aliquots of liquid from large supply bottles.
- Standard GL 45 thread on all sizes for most common bottle threads
- Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
- Screwable air vent cap for easy assembly of drying tube or microfilter
- Oval-shaped dispenser housing for easy transportation
- PFA seal of the sliding piston prevents seizing up
- Fixation slide on interior toothed track for rapid and accurate volume adjustment
- High chemical resistance of all dispensing components
- Telescopic aspirating tubes for variable adjustment to bottle height
- Fully autoclavable without disassembly
- User adjustment option
Specifications
Analog Bottletop Dispenser with Adapters | |
1 | |
2.5 to 25 mL | |
Adapters GL 32, GL 38, S 40, and telescopic intake tube (170 to 330 mm) |