Learn More
VD2-d3, MedChemExpress
MedChemExpress VD2-d3 is the deuterium labeled vitamin D2.
Brand:  Medchem Express HY-15330
Encompass_Preferred
 View more versions of this product
Product Code. 16459047
  • 3304.80 € / 5mg
Explore more special offers
Description
Description
VD2-d3 is the deuterium labeled vitamin D2.
Specifications
Specifications
VD2-d3
5 mg
Research
DMSO : 20 mg/mL (50.04 mM; ultrasonic and warming and heat to 80°C)
399.67
95.46%
Metabolism-sugar/lipid metabolism
1217448-46-8
White
C28H41D3O
O[C@@H](CCC/1=C([2H])\[2H])CC1=C([2H])\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C)C)C)([H])[C@]3(C)CCC\2
399.67
Research
Solid