missing translation for 'onlineSavingsMsg'
Learn More

VD2-d3, MedChemExpress

MedChemExpress VD2-d3 is the deuterium labeled vitamin D2.

1048.95 € - 3148.20 €

Specifications

Chemical Name or Material VD2-d3
CAS 1217448-46-8
Color White
Purity Grade Notes Research
Molecular Formula C28H41D3O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16459047
Encompass_Preferred
View Documents
Medchem Express
HY-15330
5 mg
3148.20 €
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16421509
Encompass_Preferred
View Documents
Medchem Express
HY-15330
1 mg
1048.95 €
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VD2-d3 is the deuterium labeled vitamin D2.
Specifications

Specifications

VD2-d3
White
C28H41D3O
O[C@@H](CCC/1=C([2H])\[2H])CC1=C([2H])\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C)C)C)([H])[C@]3(C)CCC\2
399.67
Research
Solid
1217448-46-8
Research
DMSO : 20 mg/mL (50.04 mM; ultrasonic and warming and heat to 80°C)
399.67
95.46%
Metabolism-sugar/lipid metabolism
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)

Recommended Storage : 4°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.