missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Powered Air Turbo Unit TR-302E+

The 3M™ Versaflo™ TR-302E+ Powered Air Turbo Unit is designed for applications involving dust and mist hazards as part of the Versaflo™ TR-300+ powered respirator system. It combines performance, protection and functionality with modern, stylish, lightweight design and unrivalled comfort.

Brand:  3M™ 7100098846

432.58 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15688566

  • 715.00 â‚¬ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Easy-to-use, versatile, powered respirator unit for pariculate environments
  • The two selectable flow rate options (185L/min and 205L/min) offer greater comfort to the user
  • Highly efficient filter captures hazardous dust, mist and fumes, plus options for nuisance level gases and vapors
  • Slim, lightweight balanced design for outstanding comfort
  • Intelligent lithium ion battery quickly charges, has no memory effect and incorporates useful charge indicator
  • Electronic audible and visual low air flow and low battery alarms as well as battery charge indicator/particulate filter loading status indicators and airflow indicator
  • Easy to adjust and maintain
  • Battery charge indicator on the battery
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Lithium-ion
Pharmaceutical manufacturing, Woodworking, Surface preparation, Food and beverage processing, Metal working, Smelting
Particulate filter, airflow indicator and spark arrestors
TR-302E+ Powered Air Turbo Unit
Versaflo™ TR-300+ motorised respirator system equipment
1135 g
Product Suggestions

Product Suggestions

Videos

Videos

  • 3M - Powered and supplied air products
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      3M Powered and supplied air products

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    3M™ Versaflo™ Powered Air Turbo Unit TR-302E+ > Product Type: TR-302E+ Powered Air Turbo Unit

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.