Learn More
Thermo Scientific™ Versette™ Pipetting Heads
Choose from a volume range of 0.5μL to 300μL with Thermo Scientific™ Versette™ Pipetting Heads.
19595.00 € - 31170.00 €
Product Code | Brand | Channels | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Channels | Volume (Metric) | Price | Quantity & Availability | ||||
15473770
|
Thermo Scientific™
650-06-384100 |
384 | 1 to 100 μL | ||||||
15198132
|
Thermo Scientific™
650-06-9630 |
96 | 0.5 to 30 μL | ||||||
15108142
|
Thermo Scientific™
650-06-96300 |
96 | 5 to 300 μL | ||||||
Description
Compatible with multiple interchangeable pipetting heads, the Thermo Scientific™ Versette™ Automated Liquid Handler provides versatile pipetting options for a broad range of applications that require 96– to 384–channel automated pipetting. The Versette system is compatible with disposable D.A.R.T.'S™, with a total volume ranging from 0.5–300μl.
The 96– and 384–channel pipetting heads use Thermo Scientific D.A.R.T.'S™, which ensure effective sealing that provides accurate and precise pipetting across all channels.
- Range from 96- to 384-channels
- All pipetting heads utilize RFID technology for self-identification.
Specifications
Pipetting Head |