missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Vessel Holder
For use with Eppendorf liquid handling products
Brand: Eppendorf™ 3113806000
This item is not returnable.
View return policy
Description
- Vessel Holder
- Includes 1 x 15mm and 1 x 24mm
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Set of 1 x 15mm and 1 x 24mm | |
Vessel Holder |