missing translation for 'onlineSavingsMsg'
Learn More

3M™ Vetbond™ Tissue Adhesive, 3 mL

Tissue Adhesive with applicator quickly and conveniently closes minor wounds to help promote healing.

Brand:  3M™ 7100139485

Product Code. 17150156

  • 403.00 € / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

3M™ Vetbond™ quickly and conveniently closes minor wounds, often eliminating the need for sutures and/or bandages. It polymerizes in seconds after contact with tissue and body fluids and binds wound edges together so natural healing can occur.

  • Strong bond, polymerizes in seconds
  • Economical 3 mL bottle with four 'reusable' applicator tips
  • Blue dye for easy-to-see drop application
  • Ideal for a variety of procedures, including: cat declaws, dew claw removal, closing minor lacerations, tooth extractions, and oral surgery
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Squeeze Bottle
3 mL (1 oz.)
Closing Minor Wounds
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Vetbond™ Tissue Adhesive, 3 mL > Tissue Adhesive; For Use With (Application): Closing Minor Wounds; Quantity: 3 mL (1 oz.); Type: Squeeze Bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.