Learn More
Bel-Art™ Vikem Vinyl Aprons
Provides protection to personnel in hospitals, schools, laboratories and industrial plants.
43.75 € - 85.00 €
Specifications
Type Apron
Size Universal
Material Vinyl
Color Clear
Disposable Reusable
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
10024610
View Documents
Bel-Art™
H24603-0001
91 cm
43.75 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
10723461
View Documents
Bel-Art™
H24604-0001
107 cm
85.00 €
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
Description
Description
  • Lightweight and comfortable
  • Made from 4mil thick vinyl
  • Stitched cotton binding securing all edges
  • Strong tie backs double reinforced with metal grommets through three layers of material at waist
  • Non-absorbent and chemically resistant
Specifications
Specifications
Apron
Vinyl
Reusable
Non-sterile
Sewn
Universal
Clear
Ties
Stitched cotton binding securing all edges
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Bel-Art™ Vikem Vinyl Aprons