missing translation for 'onlineSavingsMsg'
Learn More

Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals

24.42 € - 62.31 €

Chemical Identifiers

CAS 108-05-4
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008713
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC=C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11463890
View Documents
Thermo Scientific Alfa Aesar
A16247.AE
100 mL
24.42 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11443357
View Documents
Thermo Scientific Alfa Aesar
A16247.AP
500 mL
29.75 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11457707
View Documents
Thermo Scientific Alfa Aesar
A16247.0F
2500 mL
62.31 €
2500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.
Specifications

Chemical Identifiers

108-05-4
86.09
XTXRWKRVRITETP-UHFFFAOYSA-N
7904
ethenyl acetate
C4H6O2
MFCD00008713
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
CHEBI:46916
CC(=O)OC=C

Specifications

108-05-4
0.932
−8°C (18°F)
C4H6O2
1.396
UN1301
14,9992
Slightly miscible with water.
CC(=O)OC=C
86.09
CHEBI:46916
99%
-100°C
72°C to 73°C
Sweet
CH2=CHO2CCH3
MFCD00008713
1209327
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
XTXRWKRVRITETP-UHFFFAOYSA-N
ethenyl acetate
7904
86.09
Vinyl acetate, stabilized with 8 to 12ppm hydroquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P271-P281-P303+P361+P353-P304+P340-P308+P313-P370+P378q-P501c

H225-H332-H335-H351

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VINYL ACETATE, STABILIZED

missing translation for 'einecsNumber' : 203-545-4

missing translation for 'rtecsNumber' : AK0875000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

Product Identifier
  • Vinyl acetate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Vinyl acetate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.