Learn More
Brady™ Vinyl Cloth Labels for M611, BMP61 and BMP71
Once applied, this repositionable vinyl cloth label is there to stay...unless you want to change it.
Specifications
Autoclavable Not Autoclavable
Adhesive Type Repositionable Rubber
Color White
Finish Semi-Gloss
Material Vinyl Cloth (B-498)
View More Specs
Products 9
Product Code Brand Height (Metric) Shape Vertical Repeat (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Shape Vertical Repeat (Metric) Width (Metric) Price Quantity & Availability  
11552975
View Documents
Brady™
062668
6.4 mm Rectangle 9.5 mm 38.1 mm N/A
N/A
11582965
View Documents
Brady™
062661
7 mm Rectangle 10.2 mm 25.4 mm N/A
N/A
11552965
View Documents
Brady™
062657
12.7 mm Square 15.9 mm 12.7 mm N/A
N/A
12343478
View Documents
Brady™
062659
19.1 mm Rectangle 9.5 mm 6.4 mm N/A
N/A
11512975
View Documents
Brady™
062664
25.4 mm Rectangle 22.2 mm 19.1 mm N/A
N/A
11522975
View Documents
Brady™
062665
25.4 mm Square 28.6 mm 25.4 mm N/A
N/A
11572975
View Documents
Brady™
062670
38.1 mm Rectangle 22.2 mm 19.1 mm N/A
N/A
11542975
View Documents
Brady™
062667
101.6 mm Rectangle 104.8 mm 25.4 mm N/A
N/A
11512985
View Documents
Brady™
062674
152.4 mm Rectangle 155.6 mm 38.1 mm N/A
N/A
Description
Description
  • Adhesive and cloth backing offers excellent holding power, while allowing for clean removal and repositioning
  • Sticks to any clean, dry surface
  • Specially formulated topcoat for excellent print quality
  • Semi-gloss finish for crisp, clean look
Specifications
Specifications
Not Autoclavable
White
Vinyl Cloth (B-498)
Non-sterile
BMP™61, BMP™71, BradyPrinter M611, TLS 2200™ and TLS-PC LINKTM Thermal Labeling Systems
-40°C to +80°C
Repositionable Rubber
Semi-Gloss
Thermal Transfer
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, Wire and cable Labeling
1
0.185 mm