missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ C-Shape Vinyl Coated Lead Rings
Stabilize vessels and labware or immerse in water baths. Fisherbrand™ Vinyl-coated Lead Rings keep flasks and other labware weighted down without causing damage.
Brand: Fisherbrand™ HS8881C
35.38 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Vinyl-coated lead rings will not mark or scratch benchtops and labware
- C-shape format allows some manual size variation for a customized fit
Specifications
C-shape | |
1 Each | |
98 mm | |
0.599 kg |
Lead | |
500 to 2000 mL | |
54 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ C-Shape Vinyl Coated Lead Rings > Inner Diameter: 54mm
Spot an opportunity for improvement?Share a Content Correction