missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ C-Shape Vinyl Coated Lead Rings

Stabilize vessels and labware or immerse in water baths. Fisherbrand™ Vinyl-coated Lead Rings keep flasks and other labware weighted down without causing damage.

32.40 € - 43.00 €

Specifications

Product Type C-shape
Material Lead
Quantity 1 Each
View More Specs

Products 4
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Weight (Metric) Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Weight (Metric) Size Price Quantity & Availability  
11726725
View Documents
Fisherbrand™
HS8881A
42 mm 75 mm 0.277 kg 125 to 500 mL
32.40 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11736725
View Documents
Fisherbrand™
HS8881B
51 mm 90 mm 0.472 kg 250 to 1000 mL
39.00 €
Each
In Stock
to see stock.
Add to basket
 
11756725
View Documents
Fisherbrand™
HS8881D
66 mm 113 mm 0.907 kg 1000 to 4000 mL
43.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
 
11746725
View Documents
Fisherbrand™
HS8881C
54 mm 98 mm 0.599 kg 500 to 2000 mL
43.00 €
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
 
Description

Description

  • Vinyl-coated lead rings will not mark or scratch benchtops and labware
  • C-shape format allows some manual size variation for a customized fit
Specifications

Specifications

C-shape
Lead
1 Each
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ C-Shape Vinyl Coated Lead Rings

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.