missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Vinyl Mini Pictograms Hazardous Labels
Brand: Brady™ 814016
11.07 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
Vinyl | |
Permanent Acrylic | |
10 mm | |
-18°C to 60°C | |
Combustible Material | |
Red |
MINI PIC1803-10X10-GHS03/96 | |
96/Pk. | |
Pipe markers and safety signs | |
250 mm | |
Diamond | |
First Aid Sign |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction