missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Mini Pictograms Hazardous Labels

12.00 € - 21.58 €

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Material Vinyl
Quantity 96/Pk.
Adhesive Type Permanent Acrylic
For Use With (Application) Pipe markers and safety signs
View More Specs

Products 9
Product Code Brand Displayed Text Price Quantity & Availability  
Product Code Brand Displayed Text Price Quantity & Availability  
11313472
View Documents
Brady™
814020
CMR Mutagenic - Respiratory Risk
13.57 €
Pack of 96
In Stock
to see stock.
Edge
Add to basket
 
11353472
View Documents
Brady™
814016
Combustible Material
20.96 €
Pack of 96
In Stock
to see stock.
Edge
Add to basket
 
11333472
View Documents
Brady™
814021
Corrosive Material
21.58 €
Pack of 96
In Stock
to see stock.
Edge
Add to basket
 
12034128
View Documents
Brady™
814019
DANGER for Aquatic Environment
20.75 €
Pack of 96
In Stock
to see stock.
Edge
Add to basket
 
11303472
View Documents
Brady™
814018
DANGER of Cutaneous Sensitivity - inhalation
21.32 €
Pack of 96
In Stock
to see stock.
Edge
Add to basket
 
12024128
View Documents
Brady™
814017
Gases Under Pressure
12.00 €
Pack of 96
In Stock
to see stock.
Add to basket
 
11333442
View Documents
Brady™
814014
Explosive Material
18.33 €
Pack of 96
In Stock
to see stock.
Add to basket
 
11343442
View Documents
Brady™
814015
Inflammable Solid Material
14.92 €
Pack of 96
In Stock
to see stock.
Edge
Add to basket
 
11363472
View Documents
Brady™
814022
Toxic Material
15.50 €
Pack of 96
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Specifications

Specifications

ISO 7010, RoHS compliant using EU Directive 2002/95/EC
96/Pk.
Pipe markers and safety signs
-18°C to 60°C
Vinyl
Permanent Acrylic
10 mm
Diamond
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Vinyl Mini Pictograms Hazardous Labels

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.