missing translation for 'onlineSavingsMsg'
Learn More

3M™ Vinyl Tape 471F

Commonly used for marking hazards and aisles, color-coding, paint masking, decorative trim, splicing, and more. Made from vinyl backing with rubber adhesive, our versatile tape resists scrapes, wear, and weathering for long lasting use.

1440.00 €

Specifications

Dimensions (L x W) 50 mm x 33 m
Height (Metric) 0.1 mm
Length (Metric) 32.9 m
Thickness 0.14 mm
Width (Metric) 5.1 cm
View More Specs

Products 6
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
17207780
View Documents
3M™
7000048401
Black
1440.00 €
24 rolls
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
17257770
View Documents
3M™
7000028850
Red
1440.00 €
24 rolls
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
17267770
View Documents
3M™
7000028851
Blue
1440.00 €
24 rolls
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
17277770
View Documents
3M™
7000028852
Yellow
1440.00 €
24 rolls
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
17287770
View Documents
3M™
7000028853
White
1440.00 €
24 rolls
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
17297770
View Documents
3M™
7000028855
Green
1440.00 €
24 rolls
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
Description

Description

  • Maintains its color when exposed to abrasion, wear and many solvents
  • Unique stretch properties enable tape to remain stretched to conform to uneven surfaces without lifting and retracting
  • Ideal for use on curved and irregular surfaces
  • Easy to apply, excellent holding strength on many surfaces
  • Available in many distinctive colours (yellow, white, red, black, brown, green, orange, purple, blue and transparent)
  • Available in a wide variety of vivid colours and sizes
Specifications

Specifications

50 mm x 33 m
32.9 m
5.1 cm
0.1 mm
0.14 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.