missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Thermal Transfer Printable Labels

Printable, self-laminating vinyl labels for thermal transfer printers.

Brand:  Brady™ 236986

 View more versions of this product

Product Code. 11563145

  • 544.00 € / 1 roll

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Printable, self-laminating vinyl labels for thermal transfer printers.
  • Please Note: This material is UL recognised with its respective ribbon.
  • See the Recommended Ribbon Series for the appropriate ribbon to use.
Specifications

Specifications

ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognised
Thermal Transfer Printable Labels
White, Transparent
Vinyl
Labels
BBP11, BBP12, PR Plus, THT 76 mm Core Printers
-70°C
7000 Pk.
-94°F to +158°F
0.004 in.
38.25 mm
1.49 in.
0.5 in.
0.5 in.
BPT-305-427
Permanent Acrylic, Self-adhesive
Matte
Thermal Transfer
Data and telecommunications labelling, Electrical labelling, Laboratory labelling, Wire and cable labelling
7
7000 Labels/Roll
Rectangle
-70°C to +70°C
0.102 mm
37.85 x 12.7 mm
37.85 mm
12.7 mm
12.7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.