missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Virus Transport Medium
A ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B.
Brand: Gibco™ A48750BA
This item is not returnable.
View return policy
Description
Kit for transporting samples suspected to contain virus
- It is a ready-to-use solution containing Hanks balanced salt solution, heat inactivated FBS, gentamicin sulfate, and amphotericin B
- Suitable for the transport of samples suspected to contain virus
- Kit complies with the formulation published by the Centers for Disease Control and Prevention (CDC) for the preparation of Viral Transport Medium

Specifications
1 X | |
Virus Transport Medium | |
Sterile-filtered | |
1 L |
Liquid | |
Standard Serum Supplementation | |
No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ Virus Transport Medium > 20 L
Spot an opportunity for improvement?Share a Content Correction