Learn More
Heidolph Instruments VISCO JET™ Impeller
Ideal for mixing viscous media
Brand:  Heidolph Instruments 509-16121-00
 View more versions of this product
Product Code. 10000962
  • 121.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Useful for applications that involve viscous media such as pastes and gels that need a reliable stirring process without air entry
  • Unique blade shape triggers an intrinsically dynamic flow, which shortens processing time with significantly better mixing results compared to conventional impellers
Specifications
Specifications
10 mm shaft
170 - 300 mm Vessel
500 mm
0.30 kg
10 mm
120 mm
120 - 500 rpm
POM