missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments VISCO JET™ Impeller

Ideal for mixing viscous media

121.00 € - 501.00 €

Specifications

Diameter (Metric) Shaft 10 mm
Length (Metric) 500 mm
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Speed Material Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Speed Material Price Quantity & Availability  
10000962
View Documents
Heidolph Instruments
509-16121-00
170 - 300 mm Vessel 120 - 500 rpm POM
121.00 €
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
10356252
View Documents
Heidolph Instruments
509-16060-00
80 - 150 mm Vessel 200 to 800 rpm Stainless Steel (V4A) 501.00 €
Each
Qty Price Break Available
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
 
Description

Description

  • Useful for applications that involve viscous media such as pastes and gels that need a reliable stirring process without air entry
  • Unique blade shape triggers an intrinsically dynamic flow, which shortens processing time with significantly better mixing results compared to conventional impellers
Specifications

Specifications

10 mm
500 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.