Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Controller
The VisiLED Controllers are the core of the VisiLED system to control the VisiLED light heads.
518.00 €
Specifications
For Use With (Application) | Stereo Microscopy Illumination |
---|---|
For Use With (Equipment) | VisiLED Ringlights or Transmitted Light Stage |
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Equipment) | Price | Quantity & Availability | |||||
16637105
|
SCHOTT AG Lighting and Imaging™
400080 |
VisiLED Standard Controller MC1100 | VisiLED Ringlights or Transmitted Light Stage |
518.00 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
|||||
Description
- The compact MC1100 allows the settings of various illumination parameters including light intensity, segment modes and change of illumination direction while the MC1500 offers this for two light heads.
- Extra-fine smooth dimming.
- MC1500 offers an external trigger to start strobe function frequencies netween 15-5 KHz.
- MC1500 offers optional PC control via RS232 and footswitch control.
- Advanced setting options for optimum contrast.
- Ergonomics and blindfold operation.
- MC1500 offers control of two channels.
- Save up to four complex illumination scenarios with MC1500.
Specifications
Stereo Microscopy Illumination | |
VisiLED Ringlights or Transmitted Light Stage |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
SCHOTT AG Lighting and Imaging™ VisiLED Controller