Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Controller
The VisiLED Controllers are the core of the VisiLED system to control the VisiLED light heads.
Brand:  SCHOTT AG Lighting and Imaging™ 400080
 View more versions of this product
Product Code. 16637105
  • 518.00 € / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The compact MC1100 allows the settings of various illumination parameters including light intensity, segment modes and change of illumination direction while the MC1500 offers this for two light heads.
  • Extra-fine smooth dimming.
  • MC1500 offers an external trigger to start strobe function frequencies netween 15-5 KHz.
  • MC1500 offers optional PC control via RS232 and footswitch control.
  • Advanced setting options for optimum contrast.
  • Ergonomics and blindfold operation.
  • MC1500 offers control of two channels.
  • Save up to four complex illumination scenarios with MC1500.
Specifications
Specifications
VisiLED Standard Controller MC1100
90 x 152 x 25 mm
VisiLED Ringlights or Transmitted Light Stage
LED
CE, UKCA, EMC-class B
Stereo Microscopy Illumination
DC In, 1 Lighthead Connector, ESD plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SCHOTT AG Lighting and Imaging™ VisiLED Controller > VisiLED Controller; Type: VisiLED Standard Controller MC1100