Learn More
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Basic
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
Brand:  R&D Systems™ VCTS021
Product Code. 18772388
  • 153.00 € / 50mL
Explore more special offers
Description
Description
  • Unlike Universal solution, Acidic and Basic solutions may dramatically enhance immunoreactivity but may also affect tissue morphology.
  • The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store the unopened product at room temperature. Do not use past expiration date.
50 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Basic > 50 mL