missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ VitroEase™ Training Kit

The Thermo Scientific™ VitroEase™ Cryo-EM Training Kit provides a complete solution that includes detailed instructions for the successful preparation of grids for cryo-electron microscopy single particle analysis.

Brand:  Thermo Scientific™ A51363

815.50 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 17691546

  • 866.00 € / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The VitroEase™ Cryo-EM Training Kit has been designed to provide the necessary components to get started with cryo-EM analysis of protein samples
  • Sample preparation is a major bottleneck in cryo-electron microscopy (cryo-EM) single particle analysis (SPA)
  • Currently, sample optimization is often performed in a non-systematic way, extending optimization time over multiple days or even weeks
  • Kit includes a positive control protein, apoferritin, that has been designed specifically to meet the stringent needs for high protein purity, minimal protein aggregation, and intact protein structure
  • Commonly used accessories for grid preparation are also included such as tweezers for manipulation, grids, and grid boxes for storage
  • Detailed step-by-step instructions guide the user through the lengthy vitrification process
  • Together the control and components may be used as a comparison for success for new users or for quality control for expert users
  • Grids and accessories are provided by trusted sources
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient for 25 grid preparations.

Contains VitroEase Apoferritin Standard (Cat. No. A51362), tweezers, grids, and grid boxes.

Cryo-EM SPA
25 preparations
VitroEase™ Cryo-EM Starter Kit
Cryo-Electron Microscopes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberSPECS Certificate TypeCertificate of Analysis Date14/11/2021 Catalog NumberA51363
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ VitroEase™ Training Kit > Cryo-EM SPA

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.