missing translation for 'onlineSavingsMsg'
Learn More
Water, Deionized, Honeywell™ Product Code.: 15685340

Water, Deionized, Honeywell™

Product Code. 15685340 Shop All Honeywell Research Chemicals Products
1 L, Poly Bottle
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15685340

Brand: Honeywell Research Chemicals 387961L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Deionized

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5
Molecular Formula H2O
MDL Number MFCD00011332
UN Number NONH for all modes of transport
Beilstein 2050024
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02g/mol
Packaging Poly Bottle
Density 1.000g/mL at 3.98°C
Quantity 1 L
Chemical Name or Material Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water, Deionized, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.