missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Service Consultants™ Water Sampling Container
Sterile container with 500mL capactiy containing Sodium Thiosulphate for chlorine removal
Brand: Technical Service Consultants™ TS/15-DA40
This item is not returnable.
View return policy
Description
- 500mL Sterile Container + Sodium Thiosulphate for chlorine removal
- Please Note: the batch & expiry details are stated on the side of the label
- Shelf life of 2 years
- Chlorine level must be assessed before sampling to ensure neutralisation
Specifications
HDPE | |
500 mL | |
Neutralizing >50 mg/L Chlorine per liter of water. Ideal for water sampling of cooling towers. |
90 mg (180 mg/liter) of Sodium Thiosulphate for chlorine removal | |
Clear Bottle, Blue Cap | |
40 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction