missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™
38.20 € - 85.35 €
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.015 |
MDL Number | 11332 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | dihydrogen oxide, dihydrogen monoxide |
PubChem CID | 962 |
ChEBI | CHEBI:15377 |
IUPAC Name | oxidane |
SMILES | O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307090
|
Fisher Chemical
W/0120/PB15 |
1 L |
38.20 €
1 L |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
11357090
|
Fisher Chemical
W/0120/PB17 |
2.5 L |
85.35 €
2.50 L |
In Stock
Log in to see stock. |
|||||
Description
- Efficiency: lot-to-lot consistency ensures reproducible results
- Low acidity/alkalinity levels avoid interactions with samples
- Peak shape: low baseline noise at 210nm and 254nm improves peak shape
- Speed: filtered to 0.1μm, allowing analysis to be performed faster at higher pressures with less risk of pump blockages
- Sensitivity: interference-free with outstandingly high UV transmission for improved resolution and sensitivity
Chemical Identifiers
7732-18-5 | |
18.015 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
962 | |
oxidane |
H2O | |
11332 | |
dihydrogen oxide, dihydrogen monoxide | |
CHEBI:15377 | |
O |
Specifications
7732-18-5 | |
100°C | |
11332 | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
oxidane | |
962 | |
18.02g/mol | |
17.5mmHg at 20°C |
Colorless | |
H2O | |
dihydrogen oxide, dihydrogen monoxide | |
O | |
18.015 | |
CHEBI:15377 | |
Plastic Coated Glass Bottle | |
Liquid |
Safety and Handling
Product Identifier
- Water
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title