missing translation for 'onlineSavingsMsg'
Learn More

Lab Armor™ Waterless Bead Bath without Beads

Low maintenance and eco-friendly alternative to water baths. These Waterless Bead Baths use metallic beads instead of water, delivering exceptional temperature uniformity.

Brand:  Lab Armor™ 10122-27

Product Code. 15650034

  • 2830.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Low maintenance
  • Eco-friendly - beads are recyclable
  • Use 50% less electricity
  • No germicides needed
  • Beads hold things in place, eliminating the need for racks and weights
  • Bath is always ready, eliminating warm-up times
  • Safer operation than oil baths
  • Dry, inert, non-toxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
  • Beads have high thermal conductivity: ±180°C
  • Beads measure 5 to 8 mm dia.
Note: Beads are not included; sold separately (10122-25).

Specifications

Specifications

Waterless bead bath
20 L
Heating a variety of samples and containers, including multi-well plates, Petri dishes and open-top samples at any angle. Beads can also be used in a container for freezers, ovens and incubators.
220 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lab Armor™ Waterless Bead Bath without Beads > Waterless Bead Bath; Capacity: 20 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.