missing translation for 'onlineSavingsMsg'
Learn More

Lab Armor™ Waterless Bead Bath

Low maintenance and eco-friendly alternative to water baths. These Waterless Bead Baths use metallic beads instead of water, delivering exceptional temperature uniformity.

Brand:  Lab Armor™ 10122-05

2173.06 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15311296

  • 2630.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Low maintenance
  • Eco-friendly - beads are recyclable
  • Use 50% less electricity
  • No germicides needed
  • Beads hold things in place, eliminating the need for racks and weights
  • Bath is always ready, eliminating warm-up times
  • Safer operation than oil baths
  • Dry, inert, non-toxic aluminum oxide ensures hazard-free operation emptying, cleaning, and refilling
  • Beads have high thermal conductivity: ±180°C
  • Beads measure 5 to 8 mm dia.
Note: Beads are not included; sold separately.

Specifications

Specifications

Waterless bead bath
5 L
Heating a variety of samples and containers, including multi-well plates, Petri dishes and open-top samples at any angle. Beads can also be used in a container for freezers, ovens and incubators.
220 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers