Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand:  Watson-Marlow 981.0063.000
 View more versions of this product
Product Code. 10510176
  • 156.00 € / Each
Estimated Shipment: 01-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pump Tubing
0.63 mm
0.03 in.
323S/MC peristaltic pump
0.04 to 3.1 (four roller pump), 0.04 to 2.6 (eight roller pump)mL/min.
1/50 in.
PVC
0.8 mm
0.63mm
Orange/White