Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand:  Watson-Marlow 981.0102.000
 View more versions of this product
Product Code. 13480017
  • 74.00 € / Pack of 12
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pump Tubing
1.02 mm
0.03 in.
323S/MC peristaltic pump
0.12 to 8.1 (four roller pump), 0.09 to 6.6 (eight roller pump)mL/min.
1/25 in.
PVC
0.8 mm
1.02mm
White