Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand:  Watson-Marlow 981.0050.000
 View more versions of this product
Product Code. 17807963
  • 166.00 € / Pack of 12
Estimated Shipment: 30-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pump Tubing
0.5 mm
0.03 in.
323S/MC peristaltic pump
0.024 to 1.7 (four roller pump), 0.02 to 1.4 (eight roller pump)mL/min.
0.019 in.
PVC
0.8 mm
0.5mm
Orange/Yellow