missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ WBU-200 Series Liquid Calibration Bath

Compact and accurate WBU-200 Series Liquid Calibration Bath for external circulation or temperature calibration.

Brand:  Cole-Parmer™ 16104-50

 View more versions of this product

Product Code. 15177036

  • 1780.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fully insulated baths offer excellent heat retention.
  • Temperature stability of ±0.005°C depending on choice of control unit.
  • These compact calibration baths offer excellent stability over the entire temperature range.
  • Use these baths for external circulation to maintain temperatures of samples in viscometers, photometers, refractometers, fermenters and other reaction vessels.
  • All models offer high pump performance and exceptional thermal stability from -35°C to 250°C.
  • Add a flow cooler to the system to achieve temperatures down to -35°C.
  • The water baths are fully insulated on all sides and base and are fitted with a cooling coil for connection to a cold water supply for use at temperatures around ambient.
  • An immersion circulator is required for liquid calibration baths to operate—order Cole-Parmer IC-300 series or other compatible immersion circulators separately.
Specifications

Specifications

-35°C to +250°C
FBCAL12D
351 x 260 x 358 mm
260 mm
9 kg
Cover, lid and bridging plate
±0.005°C
351 mm
358 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.