Learn More
Tocris Bioscience™ WDR5 0103
WDR5 antagonist
Brand:  Tocris Bioscience™ 5323/50
499.34 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18761448
  • 631.00 € / 50mg
Explore more special offers
Description
Description
WDR5 0103 is a WD repeat-containing protein 5 (WDR5) antagonist (Kd = 450 nM). Disrupts WDR5 interaction with MLL and inhibits MLL core complex methyltransferase activity in vitro. Has no effect on a panel of seven other methyltransferases, including SETD7.
Specifications
Specifications
>99%
WDR5 0103
WDR5 0103
50 mg
383.44