missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WDR5 0103
WDR5 antagonist
169.00 € - 631.00 €
Specifications
Purity | >99% |
---|---|
Product Type | WDR5 0103 |
Molecular Weight (g/mol) | 383.44 |
Inhibitors | WDR5 0103 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18781348
|
Tocris Bioscience™
5323/10 |
10 mg |
169.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18761448
|
Tocris Bioscience™
5323/50 |
50 mg |
631.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WD repeat-containing protein 5 (WDR5) antagonist (Kd = 450 nM). Disrupts WDR5 interaction with MLL and inhibits MLL core complex methyltransferase activity in vitro. Has no effect on a panel of seven other methyltransferases, including SETD7.Specifications
>99% | |
WDR5 0103 | |
383.44 | |
WDR5 0103 |
For Research Use Only