Learn More
Tocris Bioscience™ WDR5 0103
WDR5 antagonist
Brand:  Tocris Bioscience™ 5323/10
133.04 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18781348
  • 169.00 € / 10mg
Explore more special offers
Description
Description
WDR5 0103 is a WD repeat-containing protein 5 (WDR5) antagonist (Kd = 450 nM). Disrupts WDR5 interaction with MLL and inhibits MLL core complex methyltransferase activity in vitro. Has no effect on a panel of seven other methyltransferases, including SETD7.
Specifications
Specifications
>99%
WDR5 0103
WDR5 0103
10 mg
383.44