Learn More
Fisherbrand™ Grade 908 Weighing Papers
Smooth paper with satin appearance and wetstrength resistance.
31.88 € - 32.10 €
Specifications
Product Type Weighing Paper
Material Paper
For Use With (Application) Weighing of all types of substances such as beet mash, powders and liquids
View More Specs
Products 2
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
11754576
View Documents
Fisherbrand™
0908F00004
150 x 150mm (5.9 x 5.9 in.)
32.10 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
11744576
View Documents
Fisherbrand™
0908F00003
95 x 110mm (3.7 x 4.3 in.)
31.88 €
Pack of 250
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Smooth paper 45g/m2
  • With satin appearance, wetstrength resistance
Specifications
Specifications
Weighing Paper
Paper
Weighing of all types of substances such as beet mash, powders and liquids
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Grade 908 Weighing Papers