Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Adapters
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
Brand:  Welch™ 828852-03
 View more versions of this product
Product Code. 10412163
  • 132.00 € / Each
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel
Welch Compact Aspiration/Fitlration Vacuum Station
Adapter, 8-Channel