Learn More
Welch™ Compact Aspiration/Filtration Vacuum Stations: Adapters
Accumulate waste liquids safe and easily. Aspiration systems prevent ingestion of fluids leading to vacuum pump failure.
132.00 €
Specifications
For Use With Welch Compact Aspiration/Fitlration Vacuum Station
View More Specs
Products 1
Product Code Brand Pump Type Material Price Quantity & Availability  
Product Code Brand Pump Type Material Price Quantity & Availability  
10412163
View Documents
Welch™
828852-03
Adapter, 8-Channel Stainless Steel
132.00 €
Each
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
Description
Description
  • Bleach resistant construction for extending service life
  • Continuous vacuum level monitoring with dial vacuum gauge
  • Powered by WOB-L piston vacuum pump
Recommended for aspirating aqueous and bleach solutions and not for acidic, basic or organic vapors and gases
Specifications
Specifications
Welch Compact Aspiration/Fitlration Vacuum Station