Learn More
Tocris Bioscience™ WH-4-023
Potent and selective Lck and Src inhibitor; also inhibits SIK
Brand:  Tocris Bioscience™ 5413/10
 View more versions of this product
Product Code. 18746509
  • 362.00 € / 10mg
Explore more special offers
Description
Description
WH-4-023 is a potent and selective Lck and Src inhibitor (IC50 values are 2 and 6 nM respectively). Exhibits >300-fold selectivity against p38α and KDR. Also potently inhibits SIK (IC50 values are 10, 22 and 60 nM for SIK 1, 2 and 3 respectively) and displays selectivity over a range of closely related kinases. Supports self-renewal of naive hESCs in combination with PD 0325901, CHIR 99021 and SB 590885 .
Specifications
Specifications
>98%
10 mg
568.67
KIN112
WH-4-023
WH-4-023