missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WH-4-023
Potent and selective Lck and Src inhibitor; also inhibits SIK
362.00 € - 1445.00 €
Specifications
Purity | >98% |
---|---|
Gene Alias | KIN112 |
Product Type | WH-4-023 |
Molecular Weight (g/mol) | 568.67 |
Inhibitors | WH-4-023 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746509
|
Tocris Bioscience™
5413/10 |
10 mg |
362.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706648
|
Tocris Bioscience™
5413/50 |
50 mg |
1445.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective Lck and Src inhibitor (IC50 values are 2 and 6 nM respectively). Exhibits >300-fold selectivity against p38α and KDR. Also potently inhibits SIK (IC50 values are 10, 22 and 60 nM for SIK 1, 2 and 3 respectively) and displays selectivity over a range of closely related kinases. Supports self-renewal of naive hESCs in combination with PD 0325901, CHIR 99021 and SB 590885 .Specifications
>98% | |
WH-4-023 | |
WH-4-023 |
KIN112 | |
568.67 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title