missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Antibiotic Assay Discs
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
225.00 € - 259.00 €
Specifications
Product Type | Antibiotic Assay Discs |
---|---|
Test Type | Infectious Disease, Antibiotic Susceptibility Sensitivity |
Packaging Type | 1000 Discs/Pk. |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11350654
|
Cytiva
2017-013 |
13 mm |
259.00 €
Pack of 1000 |
Estimated Shipment: 07-10-2024
Log in to see stock. |
|||||
11340654
|
Cytiva
2017-006 |
6 mm |
225.00 €
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
Description
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Antibiotic assay discs are used for determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.- For use with Kirby-Bauer method of antibiotic testing
- Allows rational and selective chemotherapy
Specifications
Antibiotic Assay Discs | |
Infectious Disease, Antibiotic Susceptibility Sensitivity | |
1000 Discs/Pk. |