Learn More
Cytiva Whatman™ Cellulose Extraction Thimbles
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
Brand:  Cytiva 10350240
 View more versions of this product
Product Code. 11310824
  • 109.00 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensures optimal fit with its wide range of sizes suitable for use in thimble holders and automated extraction apparatus. Whatman™ Cellulose Extraction Thimbles are constructed of high-quality cellulose for excellent mechanical strength and retention.
  • For all automated extraction apparatus
  • High-quality cellulose material
  • Dimensions matched exactly to thimble holders
Order Info
Extra allowance for wall thickness should be made when calculating external diameters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
603 Standard Cellulose Extraction Thimbles made from high-quality cellulose available in a range of sizes.
33 mm
1.5 mm
603
3.1 in.
248°F
Extraction Thimble
1.29 in.
Cellulose
For the analysis of fats or pesticides in foods and soil materials as well as in many other procedures that involve a solid-liquid extraction
For automated extraction apparatuses
80 mm
130°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ Cellulose Extraction Thimbles > Wall thickness: 1.5mm; 33 x 80mm