Learn More
QIAGEN QIAcard FTA Indicating Mini
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
Brand: QIAGEN WB120056
This item is not returnable.
View return policy
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. | |
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification,Molecular biology | |
Indicating FTA Mini Card | |
125 μL |
Buccal Cells, Saliva | |
2 | |
100 Pk. |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Acute toxicity Category 4