missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN QIAcard FTA Indicating Mini
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
185.00 € - 900.00 €
Specifications
Description | Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. |
---|---|
Sample Type | Buccal Cells, Saliva |
For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification,Molecular biology |
No. of Samples | 2 |
Product Type | Indicating FTA Mini Card |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11909434
|
QIAGEN
WB120056 |
100 Pk. |
900.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11979075
|
QIAGEN
WB120356 |
25 Pk. |
185.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
- QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
- Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
- Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards. | |
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification,Molecular biology | |
Indicating FTA Mini Card |
Buccal Cells, Saliva | |
2 | |
125 μL |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Acute toxicity Category 4