Learn More
QIAGEN QIAcard FTA Indicating Mini
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
Brand:  QIAGEN WB120356
 View more versions of this product
Product Code. 11979075
  • 185.00 € / Pack of 25
Explore more special offers
Description
Description
For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.
  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.
Specifications
Specifications
Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards.
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification,Molecular biology
Indicating FTA Mini Card
125 μL
Buccal Cells, Saliva
2
25 Pk.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning