Learn More
Cytiva Whatman™ Nylon Membranes
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
Brand: Cytiva 7402-004
Description
Flexible and durable for easier handling. Cytiva Nylon Membranes are naturally hydrophilic, eliminating need for wetting agents and prewetting when filtering aqueous solutions.
- Pure nylon membranes
- Tear-resistant
- Filter many organic solvents
- Autoclavable up to 135°C
- Biotechnology applications include filtration of tissue culture media, microbiological media, buffers and solutions
- Used in chromatography to filter samples, aqueous and organic mobile phases, vacuum degassing

Specifications
Autoclavable | |
Nylon | |
Hydrophilic | |
Filtration Membrane Disc |
Membrane Filter | |
0.2 μm | |
100/Pk. | |
47 mm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.